MAb Analysis IEX and SEC Column Kit
Thermo Scientific™  Related applications: Industrial Chromatography
Facilitate monoclonal antibody (mAb) analysis and characterization with the MAb Analysis IEX and SEC Column Kit. This kit includes an ion-exchange (IEX) and size-exclusion (SEC) column. The IEX column is intended for high-resolution and high-efficiency analysis of MAbs and their associated variants, while the SEC column is designed for separation of MAb monomers, aggregates and fragments. This kit is useful for researchers at the beginning of their MAb analysis projects.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Introducing shared lists
Back to top
Description
Facilitate monoclonal antibody (mAb) analysis and characterization with the MAb Analysis IEX and SEC Column Kit. This kit includes an ion-exchange (IEX) and size-exclusion (SEC) column. The IEX column is intended for high-resolution and high-efficiency analysis of MAbs and their associated variants, while the SEC column is designed for separation of MAb monomers, aggregates and fragments. This kit is useful for researchers at the beginning of their MAb analysis projects.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Documents
Manuals & protocols