Ammonium sulfate, 99.0-100.5%, NF, endotoxin tested, GMP, J.T.Baker™
Avantor™

Ammonium sulfate, 99.0-100.5%, NF, endotoxin tested, GMP, J.T.Baker™

This product is GMP Manufactured and meets NF specifications. Endotoxin tested.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
25 kg
50 kg
Catalog number 0798-01
Price (MXN)
-
Quantity:
500 g
This product is GMP Manufactured and meets NF specifications. Endotoxin tested.
This product is GMP Manufactured and meets NF specifications. Endotoxin tested.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS7783-20-2
Chemical Name or MaterialAmmonium sulfate
ColorWhite
Melting Point235°C
Molecular Formula(NH4)2SO4
PackagingPoly bottle, tamper-evident cap
SynonymDiammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Sulfuric acid diammonium salt
Molecular Weight (g/mol)132.14
Percent Purity99 to 100.5%
Physical FormPowder
Quantity500 g
pH5.5 (13.21 g/L, 20°C), 5 to 6 (25°C, 5% aqueous solution)
Unit SizeEach