Ammonium sulfate, 99.0-100.5%, NF, endotoxin tested, GMP, J.T.Baker™
Avantor™

Ammonium sulfate, 99.0-100.5%, NF, endotoxin tested, GMP, J.T.Baker™

This product is GMP Manufactured and meets NF specifications. Endotoxin tested.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
2.5 kg
12 kg
25 kg
50 kg
Número de catálogo 0798-F9
Precio (CLP)
-
Cantidad:
50 kg
This product is GMP Manufactured and meets NF specifications. Endotoxin tested.
This product is GMP Manufactured and meets NF specifications. Endotoxin tested.
Especificaciones
CAS7783-20-2
Nombre del producto químico o materialAmmonium sulfate
ColorBlanco
Punto de fusión235°C
Fórmula molecular(NH4)2SO4
EnvaseDrum, HDPE Flowmor package
SinónimoDiammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Sulfuric acid diammonium salt
Peso molecular (g/mol)132.14
Porcentaje de pureza99 to 100.5%
Forma físicaPowder
Cantidad50 kg
pH5 to 6 (25°C, 5% aqueous solution)
Unit SizeEach