Dionex™ IonPac™ SCS 1 Silica Cation Separator Columns
Actual product may vary
Dionex™ IonPac™ SCS 1 Silica Cation Separator Columns
Dionex™ IonPac™ SCS 1 Silica Cation Separator Columns
Thermo Scientific™

Dionex™ IonPac™ SCS 1 Silica Cation Separator Columns

Green features
For nonsuppressed conductivity detection or single-column ion chromatography (SCIC), choose this silica cation separatorcolumn.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
079809250 mm4 mmSeparator Column
079808250 mm2 mmSeparator Column
07981050 mm2 mmGuard Column
07993350 mm4 mmGuard Column
Catalog number 079809
Price (MXN)
-
Length (Metric):
250 mm
Diameter (Metric):
4 mm
Column Type:
Separator Column

For nonsuppressed conductivity detection or single-column ion chromatography (SCIC), choose the Thermo Scientific™ Dionex™ IonPac™ SCS 1 Silica Cation Separator Column. This column is particularly suited for analysis of the common inorganic cations, ammonium, select alkanolamines, and transition metals such as zinc and copper. It is the recommended column when extended calibration linearity for ammonium or alkanolamines is required.

Unique Carboxylate Cation Exchanger

The Dionex IonPac SCS 1 Silica Cation Separator column is a unique hydrophilic, low-capacity, weak cation exchanger designed for cation determinations using nonsuppressed conductivity detection. Its substrate is silica-based poly (butadiene-maleic acid) with a particle diameter of 4.5μm. The substrate bead is coated with a unique carboxylic acid functionalized layer. The Dionex IonPac SCS 1 column can be used with eluents of pH 2–7 and is compatible with typical organic solvents (such as acetonitrile and acetone) at concentration levels up to 100%. NOTE: alcohols should be avoided.

Determination of Trace Ammonium in High Sodium

The Dionex IonPac CS16 column with suppressed conductivity detection is recommended for larger concentration ratios of sodium and ammonium. The Dionex IonPac SCS 1 column can be used for regulatory compliance monitoring of alkali and alkaline earth cations and ammonium in water and wastewater.

Determination of Alkanolamines and the Common Inorganic Cations in Chemical Process Solutions

The Dionex IonPac SCS 1 column has unique selectivity for alkanolamines, and therefore can be used to resolve mixtures of these priority scrubber amines using a 3mM MSA eluent at elevated temperature.

Multiple Formats Available for Application Flexibility

  • 4x250mm, 2x250mm analytical columns
  • 4x50mm, 2x50mm guard columns
  • Smaller i.d. columns for higher mass sensitivity, eluent and reagent conservation and lower operating costs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionHydrophilic, Low-Capacity Weak Cation Exchanger Designed for Cation Determinations Using Nonsuppressed Conductivity Detection
For Use With (Application)Nonsuppressed Conductivity Detection or Single-Column Ion Chromatography (SCIC)
For Use With (Equipment)SCIC/Non-Suppressed Conductivity Detectors
Green FeaturesFSC
HydrophobicityHydrophilic
Particle Size4.5 μm
QuantityEach 1
Stationary PhaseIonPac SCS1 Silica Cation
Column TypeSeparator Column
Diameter (Metric)4 mm
FormatStandard Bore
Length (Metric)250 mm
Unit SizeEach 1