Orion™ Probe Maintenance Kits for Dissolved Oxygen Meters
Thermo Scientific™
Orion™ Probe Maintenance Kits for Dissolved Oxygen Meters
Help ensure optimal performance of dissolved oxygen probes with these specialized probe maintenance kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
080113Orion 080510MD, 081010MD, 08101 Probes
080513Orion 830A Series Probes
Catalog number 080113
Price (USD)
162.90
Online Exclusive
181.00
Save 18.10 (10%)
Each
For Use With (Equipment):
Orion 080510MD, 081010MD, 08101 Probes
Price (USD)
162.90
Online Exclusive
181.00
Save 18.10 (10%)
Each
Help ensure optimal performance of Orion polarographic dissolved oxygen probes with Thermo Scientific™ Orion™ Probe Maintenance Kits for Dissolved Oxygen Meters, which include electrolyte solution, polishing disk and two membrane caps.
Warranty: Out of box.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionFor Orion 080510MD, 081010MD, 08101 Probes
Includes2 Membrane caps, electrolyte solution, and polishing disk
For Use With (Equipment)Orion 080510MD, 081010MD, 08101 Probes
TypeProbe Maintenance Kit
Unit SizeEach