Synaptophysin Rabbit Polyclonal Antibody (clone Z66), 2ndGen Predilute

Catalog number: 081130

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This polyclonal antibody reacts with human synaptophysin. Synaptophysin (38 kDa) is a membrane component of the synaptic vessicle present in neurons and neuroendocrine cells. May be useful in identifying tumor cells of neuroendocrine origin. Positive tissue: Pancreas/neuroendocrine tumors.

Clone/PAD: Z66

Immunogen: C-terminus of human synaptophysin

Specificity: This polyclonal antibody reacts with human synaptophysin.

Reactivity: Human

Applications: Immunohistochemistry (FFPE)

Pretreatment: Epitope Retrieval: Not required, Enzyme Digestion: Not Required. Enzyme Digestion (Digest-All™ 2/Trypsin) may enhance staining.

Expected Staining Pattern: Predominantly cytoplasmic
For In Vitro Diagnostic Use.

Specifications

Reactivity: Human
Validated Application: Immunohistochemistry (FFPE)
Host Species: Rabbit
Target Gene: Synaptophysin
Gene ID: 6855
Label or Dye: Unconjugated
Dilution: 2ndGen Predilute Antibody
Product Size: 6 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols