SCLC (Small Cell Lung Cancer, CD56, N-CAM) Mouse Monoclonal Antibody (clone 123C3), 2ndGen Predilute

Catalog number: 081152

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


This monoclonal antibody reacts to the Small Cell Lung Cancer (SCLC) antigen. Clone 123C3 also reacts with the 145 and 185 kDa isomorphic glycoprotein regions of the neural cell adhesion molecule (N-CAM, CD56). The antibody stains peripheral nerve, adrenal glands, thyroid-follicular epithelium, renal tubules, hepatocytes, and pancreatic cells. It is useful in the detection of NK 8 NK-like-T-Cell lymphoma. Positive tissue: Small cell lung carcinoma

Clone/PAD: 123C3

Immunogen: SC-1 carcinoma

Specificity: SCLC (CD56)

Reactivity: Human

Applications: Immunohistochemistry (FFPE)

Pretreatment: Epitope Retrieval: Required (Citrate Buffer, pH 6.0), Enzyme Digestion: Not Required

Expected Staining Pattern: Cytoplasmic
For In Vitro Diagnostic Use.


Reactivity: Human
Validated Application: Immunohistochemistry (FFPE)
Host Species: Mouse
Target Gene: SCLC (Small Cell Lung Cancer, CD56, N-CAM)
Gene ID: 4684
Label or Dye: Unconjugated
Dilution: 2ndGen Predilute Antibody
Product Size: 6 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).


Manuals & protocols