Cytokeratin 5/6 Mouse Monoclonal Antibody (clone D5/16B4), 2ndGen Predilute

Catalog number: 081267

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This monoclonal antibody reacts with Cytokeratin 5/6, and reacts weakly with Cytokeratin 4. It stains the basal cells of squamous and glandular epithelia, part of stratum spinosum of squamous epithelium, myoepithelium, and mesothelium. It does not stain adenous epithelium. Therefore, this antibody can be used to distinguish squamous carcinoma from adenocarcinoma, and to differentiate epitheloid mesothelioma from non-epitheliod mesothelioma and adenocarcinoma. It is also a useful antibody to distinguish benign ductal hyperplasias from non-invasive neoplastic proliferations. Positive tissue: Skin.

Clone/PAD: D5/16B4

Immunogen: Purified Cytokeratin 5 and 6

Specificity: Cytokeratin 5&6. Weak reactivity with Cytokeratin 4.

Reactivity: Human

Applications: Immunohistochemistry (FFPE)

Pretreatment: Epitope Retrieval: Required (EDTA, pH 8), Enzyme Digestion: Not required

Expected Staining Pattern: Cytoplasmic
For In Vitro Diagnostic Use.

Specifications

Reactivity: Human
Validated Application: Immunohistochemistry (FFPE)
Host Species: Mouse
Target Gene: Cytokeratin 5/6
Gene ID: 140446, 3852, 3853, 3854
Label or Dye: Unconjugated
Dilution: 2ndGen Predilute Antibody
Product Size: 6 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols