Ácido túngstico(VI), Thermo Scientific Chemicals
Ácido túngstico(VI), Thermo Scientific Chemicals
Ácido túngstico(VI), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido túngstico(VI), Thermo Scientific Chemicals

CAS: 7783-03-1 | H2O4W | 249.85 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
500 g
Número de catálogo 082118.36
también denominado 082118-36
Precio (USD)
-
Cantidad:
500 g
Pedido a granel o personalizado
Identificadores químicos
CAS7783-03-1
IUPAC Namedioxotungstendiol
Molecular FormulaH2O4W
InChI KeyCMPGARWFYBADJI-UHFFFAOYSA-L
SMILESO[W](O)(=O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Impurity content1% max
Applied as flavoring agent and fragrance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only