Fill empty positions to balance the spinning rotor of the Thermo Scientific™ Rocket™ Synergy 2 Evaporator with Rocket Synergy 2 Evaporator Blanking Plugs. Order these plugs if replacements are needed.
Description | Blanking Plug - Quantity of 1 |
Type | Evaporator Blanking Plug |
For Use With (Equipment) | Rocket™ Evaporator |
Unit Size | Pack of 2 |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
082626 | Pack of 2 | Blanking Plug - Quantity of 1 | 172.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Fill empty positions to balance the spinning rotor of the Thermo Scientific™ Rocket™ Synergy 2 Evaporator with Rocket Synergy 2 Evaporator Blanking Plugs. Order these plugs if replacements are needed.