Search Thermo Fisher Scientific
Choose Thermo Scientific™ Rocket™ Synergy 2 Evaporator Lid Seals for the outer and inner lids of the Rocket Synergy 2 Evaporator if replacements are needed.
Description | Outer lid seal |
Type | Evaporator Lid Seal |
For Use With (Equipment) | Rocket Evaporator |
Unit Size | Each 1 |
Description | Inner lid seal |
Type | Evaporator Lid Seal |
For Use With (Equipment) | Rocket Evaporator |
Unit Size | Each 1 |
Description | Seal set, comprising outer and inner lid seals, and 6 flask seals |
Type | Evaporator Seal Set |
For Use With (Equipment) | Rocket Evaporator |
Unit Size | Each 1 |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
082684 | Each 1 | Outer lid seal | Price: 161.00 Online Offer: 131.40 (ends 31-Aug-2023) Your Price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
082677 | Each 1 | Inner lid seal | Price: 345.00 Online Offer: 282.60 (ends 31-Aug-2023) Your Price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
082683 | Each 1 | Seal set, comprising outer and inner lid seals, and 6 flask seals | Price: 449.00 Online Offer: 367.20 (ends 31-Aug-2023) Your Price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Save time by having your items shipped automatically.
Spend minimal time and effort to concentrate or dry large-volume samples on the Rocket Synergy 2 Evaporator for chromatographic analysis. This system provides unattended sample evaporation and with excellent reproducibility and recovery rates. Choose from several sample formats including: