Search Thermo Fisher Scientific
Choose the Thermo Scientific™ Rocket™ Synergy 2 Evaporator SampleGenie™ Flask Collar Pad and Cap if replacements are needed. The Rocket Synergy 2 Evaporator SampleGenie system allows large-volume samples to be concentrated or dried directly into a smaller storage vial on the Rocket Evaporator. An assembly, comprised of a 250 mL SampleGenie flask with a flask collar and pad, screws into the vial adapter, which holds a vial and seal. Samples in 250 mL SampleGenie flasks can be dried or concentrated into vials ranging from 12 mm to 28 mm diameter and up to 70 mm in height. Order this assembly if replacements are needed.
Description | Sample Genie Collar pad - set of 6 |
Type | Evaporator Collar Pad |
For Use With (Equipment) | Rocket Evaporator |
Unit Size | Pack of 6 |
Description | SampleGenie Collar and cap holder for GC - set of 6 |
Type | Evaporator Collar, Cap Holder |
For Use With (Equipment) | GC Vial |
Unit Size | Pack of 6 |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
082686 | Pack of 6 | Sample Genie Collar pad - set of 6 | Price: 256.00 Online Offer: 230.40 (ends 30-Nov-2023) Your Price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
082628 | Pack of 6 | SampleGenie Collar and cap holder for GC - set of 6 | Price: 2,180.00 Online Offer: 1,962.00 (ends 30-Nov-2023) Your Price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Spend minimal time and effort to concentrate or dry large-volume samples on the Rocket Synergy 2 Evaporator for chromatographic analysis. This system provides unattended sample evaporation and with excellent reproducibility and recovery rates. Choose from several sample formats including: