Thermo Scientific™
Rocket™ Synergy 2 Evaporator Flask Rack and Holder
Catalog number: 082695
Thermo Scientific™
Rocket™ Synergy 2 Evaporator Flask Rack and Holder
Catalog number: 082695
Accommodate up to six 450 mL evaporation flasks with the Thermo Scientific™ Rocket™ Synergy 2 Evaporator Flask Rack and Holder for sample filling or transport.
Catalog Number
082695
Unit Size
Each 1
Description
Flask rack, coated steel
Price (USD)
Price: 350.00
Your Price:
Availability
-
Full specifications
Quantity
DescriptionFlask rack, coated steel
MaterialSteel
TypeEvaporator Flask Rack
Holds450 mL Evaporation Flasks
Unit SizeEach 1
Catalog Number
Unit Size
Each 1
Description
Single Flask holder, for use as balance nest
Price (USD)
Price: 202.00
Your Price:
Availability
-
Full specifications
Quantity
DescriptionSingle Flask holder, for use as balance nest
TypeEvaporator Flask Holder
Holds450 mL Evaporation Flasks
Unit SizeEach 1
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)AvailabilityQuantity
082695Full specifications
Each 1Flask rack, coated steel
Price: 350.00
Your Price:
-
DescriptionFlask rack, coated steel
MaterialSteel
TypeEvaporator Flask Rack
Holds450 mL Evaporation Flasks
Unit SizeEach 1
082691Full specifications
Each 1Single Flask holder, for use as balance nest
Price: 202.00
Your Price:
-
DescriptionSingle Flask holder, for use as balance nest
TypeEvaporator Flask Holder
Holds450 mL Evaporation Flasks
Unit SizeEach 1
Showing 2 of 2
Evaporation—Pure and Simple
Spend minimal time and effort to concentrate or dry large-volume samples on the Rocket Synergy 2 Evaporator for chromatographic analysis. This system provides unattended sample evaporation and with excellent reproducibility and recovery rates. Choose from several sample formats including:
  • Thermo Scientific™ Dionex™ ASE™ Accelerated Solvent Extractor vials for convenient workflow from the Dionex ASE system to the Rocket Evaporator
  • 450 mL flasks for large-volume samples
  • Thermo Scientific™ Rocket™ Synergy 2 Evaporator SampleGenie™ vials and SampleGenie for GC autosampler vials for smaller volume samples
  • Rocket Synergy 2 Evaporator Flip-Flop vials for collecting samples in your extraction vials and transferring and evaporating them directly into GC vials
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.