Thermo Scientific™
Rocket™ Synergy 2 Evaporator Flip-Flop Vial Cover
Catalog number: 082698
Thermo Scientific™
Rocket™ Synergy 2 Evaporator Flip-Flop Vial Cover
Catalog number: 082698
Use the Thermo Scientific™ Rocket™ Synergy 2 Evaporator Flip-Flop Vial Cover if a replacement is needed for the Rocket Synergy 2 Evaporator Flip-Flop Vial. The double-ended Rocket Synergy 2 Evaporator Flip-Flop vial allows you to easily transfer your sample from a double-ended Thermo Scientific™ Dionex™ ASE™ vial into an autosampler vial. No need for robotics. Simply attach the Rocket Synergy 2 Evaporator Flip-Flop adaptor and vial to the end of the Dionex ASE collection vial and flip it over to fill the vial.
Catalog Number
082698
Unit Size
Pack of 18
Description
Flip-Flop vial cover - replacement set of 18
Price (USD)
Price: 402.00
Your Price:
Availability
-
Full specifications
Quantity
DescriptionFlip-Flop vial cover - replacement set of 18
TypeEvaporator Vial Cover
For Use With (Equipment)Rocket Evaporator Flip-Flop Vial
Unit SizePack of 18
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)AvailabilityQuantity
082698Full specifications
Pack of 18Flip-Flop vial cover - replacement set of 18
Price: 402.00
Your Price:
-
DescriptionFlip-Flop vial cover - replacement set of 18
TypeEvaporator Vial Cover
For Use With (Equipment)Rocket Evaporator Flip-Flop Vial
Unit SizePack of 18
Showing 1 of 1
Use the Thermo Scientific™ Rocket™ Synergy 2 Evaporator Flip-Flop Vial Cover if a replacement is needed for the Rocket Synergy 2 Evaporator Flip-Flop Vial. The double-ended Rocket Synergy 2 Evaporator Flip-Flop vial allows you to easily transfer your sample from a double-ended Thermo Scientific™ Dionex™ ASE™ vial into an autosampler vial. No need for robotics. Simply attach the Rocket Synergy 2 Evaporator Flip-Flop adaptor and vial to the end of the Dionex ASE collection vial and flip it over to fill the vial.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.