| 083116 | Full specifications | Each | 120 V | 22 mL System Bundle | Request A Quote |
Collection Vial Tray | 26-position tray insert for 60 mL vials; 19-position tray insert for 250 mL bottles | Depth (English) | 27.3 in. | Depth (Metric) | 69.3 cm | Description | 22 mL System Bundle | Height (English) | 24.3 in. | Height (Metric) | 61.7 cm | Pneumatic Requirements | Air at 400-827 kPa (60-120 psi); N2 (at 1034 x-1340 kPa (150-200 psi)) | Power Consumption | 500 Max VA | Weight (English) | 140 lb. | Weight (Metric) | 65 kg | Width (English) | 26.5 in. | Width (Metric) | 67.3 cm | Collection Vials | 60 mL; 250 mL | Dimensions (D x W x H) | 69.3 × 67.3 × 61.7 cm | Extraction Cell Tray | 24 Cell Positions, Two Rinse Positions, Automatic Home Position Sensing, Can Perform Multiple Extractions Per Cell | Extraction Cells | 1; 5; 10; 22; 34; 66; 100 mL | Extraction Fluids | Compatible with a wide range of organic and aqueous solvents | Fluid Sensors | IR | For Use With (Equipment) | Dionex ASE 350 | Frequency | 50/60 Hz | Pressure | 10 MPa (1500 psi) | Pump Flow | 70 mL/min | Temperature Control | Up to 200°C | Type | Dionex ASE 350 Bundle with 22 mL Cells | Voltage | 120 V | Unit Size | Each |
|
| 083114 | Full specifications | Each | 120 V | Extractor Only | Request A Quote |
Collection Vial Tray | 26-position tray insert for 60 mL vials; 19-position tray insert for 250 mL bottles | Depth (English) | 27.3 in. | Depth (Metric) | 69.3 cm | Description | Extractor Only | Height (English) | 24.3 in. | Height (Metric) | 61.7 cm | Pneumatic Requirements | Air at 400-827 kPa (60-120 psi); N2 (at 1034 x-1340 kPa (150-200 psi)) | Power Consumption | 500 Max VA | Weight (English) | 140 lb. | Weight (Metric) | 65 kg | Width (English) | 26.5 in. | Width (Metric) | 67.3 cm | Collection Vials | 60 mL; 250 mL | Dimensions (D x W x H) | 69.3 × 67.3 × 61.7 cm | Extraction Cell Tray | 24 Cell Positions, Two Rinse Positions, Automatic Home Position Sensing, Can Perform Multiple Extractions Per Cell | Extraction Cells | 1; 5; 10; 22; 34; 66; 100 mL | Extraction Fluids | Compatible with a wide range of organic and aqueous solvents | Fluid Sensors | IR | For Use With (Equipment) | Dionex ASE 350 | Frequency | 50/60 Hz | Pressure | 10 MPa (1500 psi) | Pump Flow | 70 mL/min | Temperature Control | Up to 200°C | Type | Dionex ASE 350 System | Voltage | 120 V | Unit Size | Each |
|
| 083146 | Full specifications | Each | 240 V | Extractor Only | Request A Quote |
Collection Vial Tray | 26-position tray insert for 60 mL vials; 19-position tray insert for 250 mL bottles | Depth (English) | 27.3 in. | Depth (Metric) | 69.3 cm | Description | Extractor Only | Height (English) | 24.3 in. | Height (Metric) | 61.7 cm | Pneumatic Requirements | Air at 400-827 kPa (60-120 psi); N2 (at 1034 x-1340 kPa (150-200 psi)) | Power Consumption | 500 Max VA | Weight (English) | 140 lb. | Weight (Metric) | 65 kg | Width (English) | 26.5 in. | Width (Metric) | 67.3 cm | Collection Vials | 60 mL; 250 mL | Dimensions (D x W x H) | 69.3 × 67.3 × 61.7 cm | Extraction Cell Tray | 24 Cell Positions, Two Rinse Positions, Automatic Home Position Sensing, Can Perform Multiple Extractions Per Cell | Extraction Cells | 1; 5; 10; 22; 34; 66; 100 mL | Extraction Fluids | Compatible with a wide range of organic and aqueous solvents | Fluid Sensors | IR | For Use With (Equipment) | Dionex ASE 350 | Frequency | 50/60 Hz | Pressure | 10 MPa (1500 psi) | Pump Flow | 70 mL/min | Temperature Control | Up to 200°C | Type | Dionex ASE 350 System | Voltage | 240 V | Unit Size | Each |
|
| 083115 | Full specifications | Each | 120 V | 10 mL System Bundle | Request A Quote |
Collection Vial Tray | 26-position tray insert for 60 mL vials; 19-position tray insert for 250 mL bottles | Depth (English) | 27.3 in. | Depth (Metric) | 69.3 cm | Description | 10 mL System Bundle | Height (English) | 24.3 in. | Height (Metric) | 61.7 cm | Pneumatic Requirements | Air at 400-827 kPa (60-120 psi); N2 (at 1034 x-1340 kPa (150-200 psi)) | Power Consumption | 500 Max VA | Weight (English) | 140 lb. | Weight (Metric) | 65 kg | Width (English) | 26.5 in. | Width (Metric) | 67.3 cm | Collection Vials | 60 mL; 250 mL | Dimensions (D x W x H) | 69.3 × 67.3 × 61.7 cm | Extraction Cell Tray | 24 Cell Positions, Two Rinse Positions, Automatic Home Position Sensing, Can Perform Multiple Extractions Per Cell | Extraction Cells | 1; 5; 10; 22; 34; 66; 100 mL | Extraction Fluids | Compatible with a wide range of organic and aqueous solvents | Fluid Sensors | IR | For Use With (Equipment) | Dionex ASE 350 | Frequency | 50/60 Hz | Pressure | 10 MPa (1500 psi) | Pump Flow | 70 mL/min | Temperature Control | Up to 200°C | Type | Dionex ASE 350 Bundle with 10 mL Cells | Voltage | 120 V | Unit Size | Each |
|
| 083151 | Full specifications | Each | 240 V | 10 mL System Bundle | Request A Quote |
Collection Vial Tray | 26-position tray insert for 60 mL vials; 19-position tray insert for 250 mL bottles | Depth (English) | 27.3 in. | Depth (Metric) | 69.3 cm | Description | 10 mL System Bundle | Height (English) | 24.3 in. | Height (Metric) | 61.7 cm | Pneumatic Requirements | Air at 400-827 kPa (60-120 psi); N2 (at 1034 x-1340 kPa (150-200 psi)) | Power Consumption | 500 Max VA | Weight (English) | 140 lb. | Weight (Metric) | 65 kg | Width (English) | 26.5 in. | Width (Metric) | 67.3 cm | Collection Vials | 60 mL; 250 mL | Dimensions (D x W x H) | 69.3 × 67.3 × 61.7 cm | Extraction Cell Tray | 24 Cell Positions, Two Rinse Positions, Automatic Home Position Sensing, Can Perform Multiple Extractions Per Cell | Extraction Cells | 1; 5; 10; 22; 34; 66; 100 mL | Extraction Fluids | Compatible with a wide range of organic and aqueous solvents | Fluid Sensors | IR | For Use With (Equipment) | Dionex ASE 350 | Frequency | 50/60 Hz | Pressure | 10 MPa (1500 psi) | Pump Flow | 70 mL/min | Temperature Control | Up to 200°C | Type | Dionex ASE 350 Bundle with 10 mL Cells | Voltage | 240 V | Unit Size | Each |
|
| 083152 | Full specifications | Each | 240 V | 22 mL System Bundle | Request A Quote |
Collection Vial Tray | 26-position tray insert for 60 mL vials; 19-position tray insert for 250 mL bottles | Depth (English) | 27.3 in. | Depth (Metric) | 69.3 cm | Description | 22 mL System Bundle | Height (English) | 24.3 in. | Height (Metric) | 61.7 cm | Pneumatic Requirements | Air at 400-827 kPa (60-120 psi); N2 (at 1034 x-1340 kPa (150-200 psi)) | Power Consumption | 500 Max VA | Weight (English) | 140 lb. | Weight (Metric) | 65 kg | Width (English) | 26.5 in. | Width (Metric) | 67.3 cm | Collection Vials | 60 mL; 250 mL | Dimensions (D x W x H) | 69.3 × 67.3 × 61.7 cm | Extraction Cell Tray | 24 Cell Positions, Two Rinse Positions, Automatic Home Position Sensing, Can Perform Multiple Extractions Per Cell | Extraction Cells | 1; 5; 10; 22; 34; 66; 100 mL | Extraction Fluids | Compatible with a wide range of organic and aqueous solvents | Fluid Sensors | IR | For Use With (Equipment) | Dionex ASE 350 | Frequency | 50/60 Hz | Pressure | 10 MPa (1500 psi) | Pump Flow | 70 mL/min | Temperature Control | Up to 200°C | Type | Dionex ASE 350 Bundle with 22 mL Cells | Voltage | 240 V | Unit Size | Each |
|
| 083121 | Full specifications | Each | 120 V | 34 mL System Bundle | Request A Quote |
Collection Vial Tray | 26-position tray insert for 60 mL vials; 19-position tray insert for 250 mL bottles | Depth (English) | 27.3 in. | Depth (Metric) | 69.3 cm | Description | 34 mL System Bundle | Height (English) | 24.3 in. | Height (Metric) | 61.7 cm | Pneumatic Requirements | Air at 400-827 kPa (60-120 psi); N2 (at 1034 x-1340 kPa (150-200 psi)) | Power Consumption | 500 Max VA | Weight (English) | 140 lb. | Weight (Metric) | 65 kg | Width (English) | 26.5 in. | Width (Metric) | 67.3 cm | Collection Vials | 60 mL; 250 mL | Dimensions (D x W x H) | 69.3 × 67.3 × 61.7 cm | Extraction Cell Tray | 24 Cell Positions, Two Rinse Positions, Automatic Home Position Sensing, Can Perform Multiple Extractions Per Cell | Extraction Cells | 1; 5; 10; 22; 34; 66; 100 mL | Extraction Fluids | Compatible with a wide range of organic and aqueous solvents | Fluid Sensors | IR | For Use With (Equipment) | Dionex ASE 350 | Frequency | 50/60 Hz | Pressure | 10 MPa (1500 psi) | Pump Flow | 70 mL/min | Temperature Control | Up to 200°C | Type | Dionex ASE 350 Bundle with 34 mL Cells | Voltage | 120 V | Unit Size | Each |
|
| 083153 | Full specifications | Each | 240 V | 34 mL System Bundle | Request A Quote |
Collection Vial Tray | 26-position tray insert for 60 mL vials; 19-position tray insert for 250 mL bottles | Depth (English) | 27.3 in. | Depth (Metric) | 69.3 cm | Description | 34 mL System Bundle | Height (English) | 24.3 in. | Height (Metric) | 61.7 cm | Pneumatic Requirements | Air at 400-827 kPa (60-120 psi); N2 (at 1034 x-1340 kPa (150-200 psi)) | Power Consumption | 500 Max VA | Weight (English) | 140 lb. | Weight (Metric) | 65 kg | Width (English) | 26.5 in. | Width (Metric) | 67.3 cm | Collection Vials | 60 mL; 250 mL | Dimensions (D x W x H) | 69.3 × 67.3 × 61.7 cm | Extraction Cell Tray | 24 Cell Positions, Two Rinse Positions, Automatic Home Position Sensing, Can Perform Multiple Extractions Per Cell | Extraction Cells | 1; 5; 10; 22; 34; 66; 100 mL | Extraction Fluids | Compatible with a wide range of organic and aqueous solvents | Fluid Sensors | IR | For Use With (Equipment) | Dionex ASE 350 | Frequency | 50/60 Hz | Pressure | 10 MPa (1500 psi) | Pump Flow | 70 mL/min | Temperature Control | Up to 200°C | Type | Dionex ASE 350 Bundle with 34 mL Cells | Voltage | 240 V | Unit Size | Each |
|
| 083125 | Full specifications | Each | 120 V | 66 mL System Bundle | Request A Quote |
Collection Vial Tray | 26-position tray insert for 60 mL vials; 19-position tray insert for 250 mL bottles | Depth (English) | 27.3 in. | Depth (Metric) | 69.3 cm | Description | 66 mL System Bundle | Height (English) | 24.3 in. | Height (Metric) | 61.7 cm | Pneumatic Requirements | Air at 400-827 kPa (60-120 psi); N2 (at 1034 x-1340 kPa (150-200 psi)) | Power Consumption | 500 Max VA | Weight (English) | 140 lb. | Weight (Metric) | 65 kg | Width (English) | 26.5 in. | Width (Metric) | 67.3 cm | Collection Vials | 60 mL; 250 mL | Dimensions (D x W x H) | 69.3 × 67.3 × 61.7 cm | Extraction Cell Tray | 24 Cell Positions, Two Rinse Positions, Automatic Home Position Sensing, Can Perform Multiple Extractions Per Cell | Extraction Cells | 1; 5; 10; 22; 34; 66; 100 mL | Extraction Fluids | Compatible with a wide range of organic and aqueous solvents | Fluid Sensors | IR | For Use With (Equipment) | Dionex ASE 350 | Frequency | 50/60 Hz | Pressure | 10 MPa (1500 psi) | Pump Flow | 70 mL/min | Temperature Control | Up to 200°C | Type | Dionex ASE 350 Bundle with 66 mL Cells | Voltage | 120 V | Unit Size | Each |
|
| 083156 | Full specifications | Each | 240 V | 66 mL System Bundle | Request A Quote |
Collection Vial Tray | 26-position tray insert for 60 mL vials; 19-position tray insert for 250 mL bottles | Depth (English) | 27.3 in. | Depth (Metric) | 69.3 cm | Description | 66 mL System Bundle | Height (English) | 24.3 in. | Height (Metric) | 61.7 cm | Pneumatic Requirements | Air at 400-827 kPa (60-120 psi); N2 (at 1034 x-1340 kPa (150-200 psi)) | Power Consumption | 500 Max VA | Weight (English) | 140 lb. | Weight (Metric) | 65 kg | Width (English) | 26.5 in. | Width (Metric) | 67.3 cm | Collection Vials | 60 mL; 250 mL | Dimensions (D x W x H) | 69.3 × 67.3 × 61.7 cm | Extraction Cell Tray | 24 Cell Positions, Two Rinse Positions, Automatic Home Position Sensing, Can Perform Multiple Extractions Per Cell | Extraction Cells | 1; 5; 10; 22; 34; 66; 100 mL | Extraction Fluids | Compatible with a wide range of organic and aqueous solvents | Fluid Sensors | IR | For Use With (Equipment) | Dionex ASE 350 | Frequency | 50/60 Hz | Pressure | 10 MPa (1500 psi) | Pump Flow | 70 mL/min | Temperature Control | Up to 200°C | Type | Dionex ASE 350 Bundle with 66 mL Cells | Voltage | 240 V | Unit Size | Each |
|