Dionex™ SC-CERS™ 500 Salt Converter-Cation Electrolytically Regenerated Suppressor
Actual product may vary
Thermo Scientific™
Dionex™ SC-CERS™ 500 Salt Converter-Cation Electrolytically Regenerated Suppressor
Increase sensitivity and linearity for determination of ammonium, amines and Group I and II cations using conductivity detection.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
0835492 mm
0835474 mm
Catalog number 083549
Price (USD)
2,270.00
Each 1
Description:
2 mm
Price (USD)
2,270.00
Each 1
Increase analyte response and extend linear range up to threefold by using the Thermo Scientific™ Dionex™ SC-CERS™ 500 Salt Converter-Cation Electrolytically Regenerated Suppressor to convert weakly ionized amines and ammonium to the highly ionized methanesulfonic acid (MSA) for conductivity detection. Available in a choice of 4mm format (for use with 4mm and 5mm columns) and 2mm format (for use with 2mm and 3mm columns), this simple-to-use, electrolytically regenerated suppressor requires no regenerant solution.
Enhanced Performance for Amines, Ammonium and Group I and II Cations
  • Extended linear range by up to three orders of magnitude
  • Improved sensitivity for samples with a wide range of ammonium or amine concentrations
  • Reduced number of calibration check standards needed because of its improved accuracy at higher concentrations
  • Increased sample throughput for industries with high sample workloads, such as the power generation industry
  • Updated design with increased backpressure tolerance and maximum leak tolerance
  • Lowered noise and fast startup thanks to innovative regenerant sweep-out design
  • Saves time with self-regenerating functionality
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for six (6) months thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description2 mm
For Use With (Application)Conductivity Detection
OperationConvert Analytes to Fully-Dissociated Methanesulfonic Acid Species
Void Volume<15 μL
TypeSalt Converter-Cation Electrolytically Regenerated Suppressor
Unit SizeEach 1