B·R·A·H·M·S™ CgA II KRYPTOR™
Thermo Scientific™
B·R·A·H·M·S™ CgA II KRYPTOR™
Thermo Scientific B·R·A·H·M·S CgA II KRYPTOR is the first-FDA-cleared, automated immunofluorescent assay for determining CgA concentration in human serum.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDescripciónPara utilizar con (aplicación)
083991NCalibrador para B·R·A·H·M·S CgA II KRYPTORReajustar la curva estándar memorizada en B·R·A·H·M·SKRYPTOR compact PLUS / KRYPTOR GOLD
839050NEnsayo inmunofluorescente para la determinación cuantitativa de cromogranina A soluble (CgA)La prueba B·R·A·H·M·S CgA II KRYPTOR está indicada como ayuda para su uso junto con la evaluación clínica para el seguimiento y la monitorización de pacientes con tumores neuroendocrinos (TNE) y para la monitorización del tratamiento de pacientes con cáncer de próstata. La prueba también está indicada como ayuda para su uso junto con la evaluación clínica para el diagnóstico de pacientes con sospecha de feocromocitomas. Consulte las instrucciones de uso disponibles en su país.
083992NControles para B·R·A·H·M·S CgA II KRYPTOR (2 niveles)Control de calidad del ensayo B·R·A·H·M·S CgA II KRYPTOR
Número de catálogo 083991N
Precio (MXN)
-
Descripción:
Calibrador para B·R·A·H·M·S CgA II KRYPTOR
Para utilizar con (aplicación):
Reajustar la curva estándar memorizada en B·R·A·H·M·SKRYPTOR compact PLUS / KRYPTOR GOLD
Thermo Scientific™ B·R·A·H·M·S™ CgA II KRYPTOR™ is an automated immunofluorescent assay using Time-Resolved amplified Cryptate Emission (TRACE™) technology to determine the concentration of Chromogranin A (CgA) in human serum. B·R·A·H·M·S CgA II KRYPTOR is indicated as an aid to be used in conjunction with clinical evaluation for follow-up and monitoring of patients with neuroendocrine tumors (NETs) and for therapy monitoring of patients with prostate cancer. The test is further indicated as an aid to be used in conjunction with clinical evaluation for the diagnosis of patients with suspected pheochromocytomas. Please consult the Instructions For Use (IFU) available in your country.
Intended Use for US only: B·R·A·H·M·S CgA II KRYPTOR is to be used in conjunction with other clinical methods as an aid in monitoring of disease progression during the course of disease and treatment in patients with gastroentero-pancreatic neuroendocrine tumors (GEP-NETs, grade 1 and grade 2).
  • Chromogranin A is an acidic, hydrophilic protein of 49 kDa present in chromaffin granules of the neuroendocrine cells and is a member of the granin family.
  • Neuroendocrine tumors (NETs) originate from neuroendocrine cells found in neuronal and endocrine tissues throughout the body.
  • The most common sites of NETs are the lung, stomach, appendix, cecum, duodenum, pancreas, jejunum/ileum, colon and rectum.
  • NETs arising from the gastrointestinal tract are collectively known as gastroentero-pancreatic neuroendocrine tumors.
  • Follow up and monitoring investigations of NETs are based on tumor markers together with other diagnostic measures and clinical signs.
  • Besides its use as immunohistological marker, CgA has been well recognized as a general broad-spectrum serum marker in GEP-NETs.
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
Especificaciones
Tiempo de análisis29 min.
DescripciónCalibrador para B·R·A·H·M·S CgA II KRYPTOR
Para utilizar con (aplicación)Reajustar la curva estándar memorizada en B·R·A·H·M·SKRYPTOR compact PLUS / KRYPTOR GOLD
IncluyeCalibrator kit, 6 vials
Analitos detectablesChromogranin A (CgA)
FormularioLyophilized
Unit SizePack of 6
Contenido y almacenamiento
Calibrator kit, 6 vials, 2-8°C