B·R·A·H·M·S™ hCG+β KRYPTOR™
Thermo Scientific™
B·R·A·H·M·S™ hCG+β KRYPTOR™
B·R·A·H·M·S provides a complete portfolio of biomarkers for the most reliable results in prenatal screening. Get the quadruple test for trisomy screening in second trimester of pregnancy with B·R·A·H·M·S hCG+β KRYPTOR in less than 40 minutes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
084193NControl (1 Level High)
841050NImmunofluorescent Assay
084191NCalibrator
084192NControls (2 Levels)
Catalog number 084193N
Price (EUR)
-
Type:
Control (1 Level High)
B·R·A·H·M·S provides a complete portfolio of biomarkers for the screening of chromosomal abnormalities in first and second trimester of pregnancy.B·R·A·H·M·S hCG + β KRYPTOR is indicated as an aid for risk assessment of trisomy 21 and 18 in the second trimester.
US Only: Product is not 510k cleared in US.
Get confidence in your results with the high precision of B·R·A·H·M·S hCG+β KRYPTOR and its short incubation time.
  • Incubation time of only 14 minutes for a short turnaround time
  • Allows measurement in serum
  • Large capacity with 50 tests
  • Kit stable up to 29 days after opening onboard the instrument
  • Easy workflow with only 1 calibrator and one calibration every 2 weeks
  • Fits your throughput need with B·R·A·H·M·S KRYPTOR compact PLUS or B·R·A·H·M·S KRYPTOR GOLD
  • Allows risk calculation in second trimester with B·R·A·H·M·S Fast screen pre I plus
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
Specifications
Analysis Time14 minutes
For Use With (Application)Quality Control for B·R·A·H·M·S hCG+β KRYPTOR assay
For Use With (Equipment)B·R·A·H·M·S hCG+β KRYPTOR
IncludesControls kit, 6 vials
TypeControl (1 Level High)
Detectable Analytesbeta-hCG
FormLyophilized
Unit SizeBox with 6 vials
Contents & Storage
Controls kit, 6 vials, 2°C to 8°C