MAbPac™ SCX-10 HPLC Columns
Thermo Scientific™

MAbPac™ SCX-10 HPLC Columns

Green features

Use these strong cation-exchange columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle SizeColumn Format
085198150 mm4 mm5 μmAnalytical Column
074625250 mm4 mm10 μmAnalytical Column
075602150 mm4 mm10 μmAnalytical Column
SP6947250 mm22 mm10 μmPreparative Column, Standard
075603TS
also known as 075603
50 mm4 mm10 μmAnalytical Column
075604250 mm2 mm10 μmAnalytical Column
08267450 mm4.6 mm5 μmAnalytical Column
085209150 mm4.6 mm5 μmAnalytical Column
082673250 mm4.6 mm5 μmAnalytical Column
078655250 mm4 mm5 μmAnalytical Column
07865650 mm4 mm5 μmAnalytical Column
07574950 mm2 mm10 μmGuard Column
07463150 mm4 mm10 μmGuard Column
088784250 mm9 mm10 μmPreparative Column, Standard
088783250 mm4 mm10 μmAnalytical Column Set (3 Columns, 3 Lots)
Catalog number 085198
Price (USD)
2,083.63
Each
Length (Metric):
150 mm
Diameter (Metric):
4 mm
Particle Size:
5 μm
Column Format:
Analytical Column
Price (USD)
2,083.63
Each

Use Thermo Scientific™ MAbPac™ SCX-10 LC Columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants. The unique nonporous pellicular resin provides the resolving power to separate mAb variants that differ by as little as one charged residue. A hydrophilic layer surrounds the polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.

Superior Performance for mAb Variant Analysis
  • Exceptionally high resolution for mAb variants
  • Ideal for characterization and quality control assessment of mAbs
  • Well-suited for stability studies, mAb therapeutic method development and high-throughput screening
  • Unmatched column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
  • Meets the regulatory requirements for biopharmaceutical characterization
Multiple Sizes Available
  • Available in 3, 5, and 10μm particle sizes in standard and UHPLC formats
  • Smaller-particle-size columns provide the same resolution as the 10μm columns, but with significantly faster analysis time
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeIon Exchange
Green FeaturesFSC, ACT Ecolabel (Gold)
Max. Pressure5000 psi (345 bar)
Packing MaterialSpherical, Nonporous Polymer
Product TypeAnalytical Column
QuantityEach
Stationary PhaseSulfonate
TemperatureAmbient to 60°C
Diameter (Metric)4 mm
Length (Metric)150 mm
Particle Size5 μm
PhaseIon Exchange
Pore SizeNonporous
Product LineMAbPac
TechniqueBio-LC
pH2 to 12
Unit SizeEach