Product Image
Thermo Scientific™
Orion™ RDO Replacement Electrode Cap
Use the probe optical cap replacement with RDO probes.
Have Questions?
Catalog NumberType
087001Probe Optical Cap
Catalog number 087001
Price (USD)
155.70
Online exclusive
173.00
Save 17.30
Each
Type:
Probe Optical Cap
Price (USD)
155.70
Online exclusive
173.00
Save 17.30
Each
Use the Thermo Scientific™ Orion™ RDO™ Probe Optical Cap Replacement with Orion RDO probes.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Do not use solvents or soaps to clean the RDO cap and do not rub the cap with abrasive material. Use of these materials will damage the RDO cap and void the warranty. Do not let salt or mineral deposits form on the RDO cap.
Specifications
DescriptionOptical Cap
For Use With (Equipment)Orion RDO Probes 087010MD, 087020MD, 087030MD, 087050MD, 087100MD
TypeProbe Optical Cap
Unit SizeEach