CTS™ AIM V™ is now available without phenol red and antibiotics (A3830801 and A3830802).
CTS™ AIM V™ SFM
CTS™ AIM V™ SFM
CTS™ AIM V™ SFM
Gibco™

CTS™ AIM V™ SFM

CTS AIM V SFM is a defined, serum-free medium (SFM) for the proliferation and/or manipulation of T cells, dendritic cells, and other immune cells manufactured in compliance with cGMP.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
0870112BK10 L
0870112DK1 L
Catalog number 0870112BK
Price (USD)
1,760.00
Each
Quantity:
10 L
Price (USD)
1,760.00
Each

Gibco Cell Therapy Systems (CTS) AIM-V SFM is a defined, serum-free medium specifically designed for the proliferation and/or manipulation of T cells, dendritic cells, and other immune cells that is manufactured in compliance with current Good Manufacturing Practice (cGMP) standards.

CTS AIM V SFM contains L-glutamine, 50 μg/mL streptomycin sulfate, and 10 μg/mL gentamicin sulfate.

CTS quality

Gibco CTS products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485-certified. Gibco CTS products follow USP 'ancillary materials for cell, gene, and tissue-engineered products' within the responsibilities applicable to a supplier.*

Accompanied by documentation to support your regulatory filing

Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available.

*Other aspects of USP will be the responsibility of the end-user to assess. Thermo Fisher Scientific cannot fulfill USP in regards to application and therapy-specific aspects (e.g., residual assessment and removal of the Ancillary Material).

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Cell TypeMonocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
FormatBPC
Manufacturing QualityISO 13485, MDSAP, FDA-registered, 21 CFR 820
Product TypeCell Culture Medium
Quantity10 L
Shelf Life14 months from date of manufacture
Shipping ConditionAmbient
ClassificationAnimal Origin
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
Unit SizeEach
Contents & Storage
Store at 2–8°C and protect from light.

Frequently asked questions (FAQs)

What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

Will CTS AIM V SFM (Cat. No. 0870112BK) be discontinued?

No. CTS AIM V SFM (Cat. No. 0870112BK) will not be discontinued. The same catalog number will be available with the new Aegis5-14 film following the planned transition in 2026. 

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Where can I get more information about Aegis 5-14 BPC (BioProcessing Container)?

For additional information regarding Aegis 5-14 BPC, reach out to our Cell Culture Technical Support Team at techsupport@thermofisher.com. 

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What type of film is currently used for packaging CTS AIM V SFM (Cat. No. 0870112BK)?

CTS AIM V SFM (Cat. No. 0870112BK) is packaged in ASI 28 film, featuring an ethylene vinyl acetate (EVA) contact layer and four spike ports.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between CTS AIM V SFM VALIDATION (Cat. No. A4000311302) and CTS AIM V SFM (Cat. No. 0870112BK)?

The two media have the same formulation and GMP quality and are manufactured in ISO13485, MDSAP, FDA-registered, 21 CFR820 facilities. The main difference lies in the packaging. CTS AIM V SFM VALIDATION (Cat. No. A4000311302) is packaged in the Aegis 5-14 BioProcessing Container (BPC) with a low-density polyethylene (LDPE) contact layer and updated tubing and connector options. On the other hand, the current CTS AIM V SFM (Cat. No. 0870112BK) is packaged in ASI 28 film with an ethylene vinyl acetate (EVA) contact layer and four spike ports.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (27)

Citations & References
Abstract
Live attenuated lentivirus infection elicits polyfunctional simian immunodeficiency virus Gag-specific CD8+ T cells with reduced apoptotic susceptibility in rhesus macaques that control virus replication after challenge with pathogenic SIVmac239.
Authors:Genescà M, Rourke T, Li J, Bost K, Chohan B, McChesney MB, Miller CJ,
Journal:J Immunol
PubMed ID:17878372
'HIV-specific CD8+ T cells that secrete multiple cytokines in response to Ag stimulation are associated with the control of virus replication during chronic HIV infection. To determine whether the presence of polyfunctional CD8+ T cell responses distinguishes protected and unprotected monkeys in a live attenuated lentivirus model, SIV Gag peptide-specific ... More
Performance of serum-supplemented and serum-free media in IFNgamma Elispot Assays for human T cells.
Authors:Janetzki S, Price L, Britten CM, van der Burg SH, Caterini J, Currier JR, Ferrari G, Gouttefangeas C, Hayes P, Kaempgen E, Lennerz V, Nihlmark K, Souza V, Hoos A,
Journal:Cancer Immunol Immunother
PubMed ID:19894047
'The choice of serum for supplementation of media for T cell assays and in particular, Elispot has been a major challenge for assay performance, standardization, optimization, and reproducibility. The Assay Working Group of the Cancer Vaccine Consortium (CVC-CRI) has recently identified the choice of serum to be the leading cause ... More
[Study of adoptive immunotherapy for metastatic renal cell carcinoma with lymphokine-activated killer (LAK) cells and interleukin-2. II. Clinical evaluation]
Authors:Nomura K, Fujioka T,
Journal:Nippon Hinyokika Gakkai Zasshi
PubMed ID:8320888
'We evaluated adoptive immunotherapy using LAK cells combined with systemic administration of interleukin-2 (IL-2) in 11 patients with metastatic renal cell carcinoma. The LAK cells were generated by incubation in serum-free medium (AIM-V) supplemented with IL-2 (1,000 U/ml) for 4 days and were generally administered twice weekly (4 times/cycle). Daily ... More
Abnormal acidification of melanoma cells induces tyrosinase retention in the early secretory pathway.
Authors: Halaban Ruth; Patton Robin S; Cheng Elaine; Svedine Sherri; Trombetta E Sergio; Wahl Miriam L; Ariyan Stephen; Hebert Daniel N;
Journal:J Biol Chem
PubMed ID:11812790
'In tyrosinase-positive amelanotic melanoma cells, inactive tyrosinase accumulates in the endoplasmic reticulum. Based on studies described here, we propose that aberrant vacuolar proton ATPase (V-ATPase)-mediated proton transport in melanoma cells disrupts tyrosinase trafficking through the secretory pathway. Amelanotic but not melanotic melanoma cells or normal melanocytes display elevated proton export ... More
Serum-free culture medium and IL-7 costimulation increase the sensitivity of ELISpot detection.
Authors:Martinuzzi E, Scotto M, Enée E, Brezar V, Ribeil JA, van Endert P, Mallone R
Journal:J Immunol Methods
PubMed ID:18242633
'The identification of parameters maximizing detection sensitivity in ELISpot assays is important to transfer this technology into the clinical setting for identifying rare Ag-specific CD8(+) T cells. We have therefore considered human IFN-gamma CD8(+) T cell responses against viral epitopes to analyze different variables which could be critical during the ... More