Gibco CTS AIM V SFM (therapeutischer Gütegrad) ist das erste im Handel erhältliche definierte, serumfreie Medium für die Proliferation und/oderWeitere Informationen
Have Questions?
Ansicht ändern
Katalognummer
Menge
0870112DK
1.000 mL
0870112BK
10 l
2 Optionen
Katalognummer 0870112DK
Preis (EUR)
302,00
Each
Menge:
1.000 mL
Preis (EUR)
302,00
Each
Gibco CTS AIM V SFM (therapeutischer Gütegrad) ist das erste im Handel erhältliche definierte, serumfreie Medium für die Proliferation und/oder Manipulation von T-Zellen und dendritischen Zellen, das gemäß cGMP hergestellt wird. Es enthält L-Glutamin, 50 µg/ml Streptomycin-Sulfat und 10 µg/ml Gentamicin-Sulfat.
Für Anwendungen zur Verarbeitung von humanen Ex-vivo-Geweben und Zellkulturen.ACHTUNG:Für medizinische Anwendungen darf dieses Produkt gemäß US-amerikanischer Bundesgesetzgebung nur von einem Arzt oder auf dessen Anweisung erworben werden.
Medium bei 2 bis 8°C lagern. Vor Licht schützen. AIM V™ hat eine Haltbarkeit von 14 Monaten.
Häufig gestellte Fragen (FAQ)
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
What is the difference between AIM V Medium (Cat. Nos. 12055083, 12055091) and CTS AIM V SFM (Cat. Nos. 0870112DK, 0870112BK)?
The difference is in the intended usage of the two media. AIM V Medium (Cat. Nos. 12055083, 12055091) is for research use only whereas CTS AIM V SFM (Cat. Nos. 0870112DK, 0870112BK) is for research use or manufacturing of cell, gene, or tissue-based products.
What is the difference between CTS AIM-V Medium and CTS AIM V SFM?
These two media have comparable performance and same cGMP quality. The main difference is that CTS AIM-V Medium (Cat. Nos. A3830801, A3830802, A4672701) does not contain phenol red and antibiotics. This medium formulation has been used as an ancillary reagent in a therapeutic cancer vaccine approved by FDA.
Live attenuated lentivirus infection elicits polyfunctional simian immunodeficiency virus Gag-specific CD8+ T cells with reduced apoptotic susceptibility in rhesus macaques that control virus replication after challenge with pathogenic SIVmac239.
Authors:Genescà M, Rourke T, Li J, Bost K, Chohan B, McChesney MB, Miller CJ,
Journal:J Immunol
PubMed ID:17878372
'HIV-specific CD8+ T cells that secrete multiple cytokines in response to Ag stimulation are associated with the control of virus replication during chronic HIV infection. To determine whether the presence of polyfunctional CD8+ T cell responses distinguishes protected and unprotected monkeys in a live attenuated lentivirus model, SIV Gag peptide-specific ... More
Performance of serum-supplemented and serum-free media in IFNgamma Elispot Assays for human T cells.
Authors:Janetzki S, Price L, Britten CM, van der Burg SH, Caterini J, Currier JR, Ferrari G, Gouttefangeas C, Hayes P, Kaempgen E, Lennerz V, Nihlmark K, Souza V, Hoos A,
Journal:Cancer Immunol Immunother
PubMed ID:19894047
'The choice of serum for supplementation of media for T cell assays and in particular, Elispot has been a major challenge for assay performance, standardization, optimization, and reproducibility. The Assay Working Group of the Cancer Vaccine Consortium (CVC-CRI) has recently identified the choice of serum to be the leading cause ... More
[Study of adoptive immunotherapy for metastatic renal cell carcinoma with lymphokine-activated killer (LAK) cells and interleukin-2. II. Clinical evaluation]
Authors:Nomura K, Fujioka T,
Journal:Nippon Hinyokika Gakkai Zasshi
PubMed ID:8320888
'We evaluated adoptive immunotherapy using LAK cells combined with systemic administration of interleukin-2 (IL-2) in 11 patients with metastatic renal cell carcinoma. The LAK cells were generated by incubation in serum-free medium (AIM-V) supplemented with IL-2 (1,000 U/ml) for 4 days and were generally administered twice weekly (4 times/cycle). Daily ... More
Serum-free culture medium and IL-7 costimulation increase the sensitivity of ELISpot detection.
Authors:Martinuzzi E, Scotto M, Enée E, Brezar V, Ribeil JA, van Endert P, Mallone R
Journal:J Immunol Methods
PubMed ID:18242633
'The identification of parameters maximizing detection sensitivity in ELISpot assays is important to transfer this technology into the clinical setting for identifying rare Ag-specific CD8(+) T cells. We have therefore considered human IFN-gamma CD8(+) T cell responses against viral epitopes to analyze different variables which could be critical during the ... More
CD4 T cells guarantee optimal competitive fitness of CD8 memory T cells.
Authors:Johansen P, Stamou P, Tascon RE, Lowrie DB, Stockinger B,
Journal:Eur J Immunol
PubMed ID:14971034
We studied the contribution of CD4 T cell help to survival and competitive fitness of CD8 memory T cells specific for influenza virus nucleoprotein. In agreement with recent studies, the optimal generation of functional memory CD8 T cells required CD4 help, although long-term maintenance of resting CD8 memory T cells ... More