Phenylmagnesium bromide, 3M in ether
Phenylmagnesium bromide, 3M in ether
Phenylmagnesium bromide, 3M in ether
Thermo Scientific Chemicals

Phenylmagnesium bromide, 3M in ether

Grignard reagent | CAS: 100-58-3 | C6H5BrMg | 181.32 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
500 mL
產品號碼 087326.AP
亦稱為 087326-AP
價格 (TWD)
-
Quantity:
500 mL
Request bulk or custom format
化學識別
CAS100-58-3
IUPAC Namebromo(phenyl)magnesium
Molecular FormulaC6H5BrMg
InChI KeyANRQGKOBLBYXFM-UHFFFAOYSA-M
SMILESBr[Mg]C1=CC=CC=C1
檢視更多
規格Specification Sheet規格表
Assay (Titration)2.7 to 3.3 M
FormClear liquid
Grignard reagent Phenylmagnesium bromide is an important reagent called as Grignard reagent used in the synthesis of organic and oraganometallic reactions. It is used to prepare phenyl aryl sulphides, sulphoxides, and sulphones. It acts as a strong acid and Lewis acid and undergoes addition reaction with many unsaturated functional groups. It is involved in the synthesis of end-functionalized regioregular poly(3-alkylthiophene)s, 1,3,3-trimethyl-6-phenyl-2-oxabicyclo[2.2.2]octan-6-ol, o-substituted benzophenones and 6-benzyl-1,3,3-trimethyl-2-oxabicyclo[2.2.2]octan-6-ol. In addition to this, it is used for the monalkylation of primary amines to get secondary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only