Iodic acid, ACS, 99.5% min
Iodic acid, ACS, 99.5% min
Iodic acid, ACS, 99.5% min
Thermo Scientific Chemicals

Iodic acid, ACS, 99.5% min

CAS: 7782-68-5 | HIO3 | 175.91 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
50 g
250 g
產品號碼 087681.30
亦稱為 087681-30
價格 (TWD)
-
Quantity:
250 g
Request bulk or custom format
化學識別
CAS7782-68-5
IUPAC Nameiodic acid
Molecular FormulaHIO3
InChI KeyICIWUVCWSCSTAQ-UHFFFAOYSA-N
SMILESO[I](=O)=O
檢視更多
規格Specification Sheet規格表
Impurity contentIodide P.T.
Impurity contentNitrogen compounds (as N) 0.1%
Impurity contentSO₄ 0.015%
Assay (unspecified)99.5% HIO₃ min
Residue on ignition0.02%
檢視更多
Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only