Thermo Scientific™

Molecular sieves, 4A, 1-2mm (0.04-0.08in) dia. pellets, Thermo Scientific™

5kg, Each
Thermo Scientific™

Molecular sieves, 4A, 1-2mm (0.04-0.08in) dia. pellets, Thermo Scientific™

5kg, Each
Quantity
Catalog number: 087956.A7
also known as 087956-A7
Price (USD)
348.00 / Each
Your Price:
Quantity
-

Chemical Identifiers

CAS
70955-01-0
Appearance (Color)
White to off-white
Form
1 to 2mm pellets
Assay (unspecified)
> 21 wt% water absorption capacity
Loss on ignition
1.5%, max

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only

Figures