Thallium(III) nitrate trihydrate, 99.5% (metals basis)
Thallium(III) nitrate trihydrate, 99.5% (metals basis)
Thallium(III) nitrate trihydrate, 99.5% (metals basis)
Thermo Scientific Chemicals
Thallium(III) nitrate trihydrate, 99.5% (metals basis)
CAS: 13453-38-8 | H12N3O15Tl | 498.48 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 087999.14
または、製品番号087999-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13453-38-8
Total Metal Impurities0.5% max.
Thallium(III) nitrate trihydrate is a reagent used for the rapid oxidation of styrene derivatives to carbonyl compounds, for the regioselective ring expansion of cyclic aralkyl ketones via oxidative rearrangement, for the selective hydrolysis of thioacetals to carbonyl compounds and for a general synthetic method to 19-norsteroids. Mediates ring contraction of cyclic ketones to a series of trans-fused 10-methyl-2-decalones.