Platinum, AAS standard solution, Specpure™ Pt 1000μg/mL
Platinum, AAS standard solution, Specpure™ Pt 1000μg/mL
Platinum, AAS standard solution, Specpure™ Pt 1000μg/mL
Thermo Scientific Chemicals

Platinum, AAS standard solution, Specpure™ Pt 1000μg/mL

Form: Liquid | CAS: 7440-6-6 | Pt in 20% HCl
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 088086.AE
also known as 088086-AE
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
Assay Percent RangePt 1000μg/mL
CAS6-6-7440
Chemical Name or MaterialPlatinum
DOT InformationDOT Class: 8, Packaging Group: II
DensityApprox. 1g/cm3 at 20°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only