Specifications
Chemical Name or MaterialSamarium, AAS standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECS Number231-714-2
Health Hazard 1H290-H314-H332-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
View more