GlycanPac™ AXR-1 HPLC Columns
Thermo Scientific™

GlycanPac™ AXR-1 HPLC Columns

Green features

Get ultra-high resolution and selectivity for both labeled and native glycans using this HPLC/UHPLC column. It's compatible with MS and fluorescence detection.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
088135250 mm2.1 mm1.9 μm
088136150 mm2.1 mm1.9 μm
Catalog number 088135
Price (USD)
1,800.08
Each
Length (Metric):
250 mm
Diameter (Metric):
2.1 mm
Particle Size:
1.9 μm
Price (USD)
1,800.08
Each

Get industry-leading, ultra-high resolution of labeled and native glycans for structural characterization and quantification using the Thermo Scientific™ GlycanPac™ AXR-1 LC Columns. This high-performance, silica-based chromatography column achieves mixed-mode separation using a combination of charge, isomer structure, and size. Now there's a column with unique selectivity to analyze biologically important glycans. The GlycanPac AXR-1 column is available in multiple HPLC and UHPLC formats and is compatible with fluorescence or mass spectrometry (MS) detection.

Exceptional Glycan Selectivity with Industry-Leading Resolution

Ultra-high resolution and selectivity of glycans are attributed to the GlycanPac AXR-1 column's innovative mixed-mode surface chemistry, which combines:

  • WAX functionality for retention of negatively charged glycans, with separation in order of increasing charge
  • Reversed-phase functionality for same-charge glycans, with separation according to isomerism and size
Reliable Column Performance

This high-purity spherical silica column is designed and tested for HPLC and UHPLC methods using either fluorescence or MS detection applications. The GlycanPac AXR-1 column uses volatile aqueous buffers (e.g., ammonium acetate or ammonium formate) and acetonitrile, presenting the eluting glycans ready for introduction into MS instruments.

Available in Multiple Formats
  • Use 1.9 μm columns for UHPLC applications and 3 μm columns for HPLC applications
  • Select from a variety of dimensions to support the resolution, throughput, or instrumental requirements required for your application
  • Protect the analytical column with GycanPac AXR-1 Guard columns
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column ChemistryWAX and RP Mixed Mode
Column FormatAnalytical Column
Column TypeMixed Mode
Flow Rate0.2 to 0.4 mL/min
Green FeaturesFSC
Max. Pressure15000 psi (1035 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Particle ShapeSpherical, high purity, porous
Product TypeAnalytical Column
QuantityEach
SolventsCompatible with 0 to 100% aqueous and common HPLC solvents
Stationary PhaseProprietary
Surface Area220 m²/g
Temperature60°C
Diameter (Metric)2.1 mm
Length (Metric)250 mm
Particle Size1.9 μm
PhaseReversed Phase and Ion Exchange
Pore Size175 Å
Product LineGlycanPac
TechniqueBio-LC
pH2 to 8
Unit SizeEach