Thermo Scientific Chemicals

Strontium oxide, 99.5% (metals basis), SrO ^=97%, Thermo Scientific Chemicals

Catalog number: 088220.22
100 g, Each
Thermo Scientific Chemicals

Strontium oxide, 99.5% (metals basis), SrO ^=97%, Thermo Scientific Chemicals

Catalog number: 088220.22
100 g, Each
Quantity
Catalog number: 088220.22
also known as 088220-22
Price (USD)
Price: 425.00
Online price: 361.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1314-11-0
IUPAC Name
strontium(2+) oxidandiide
Molecular Formula
OSr
InChI Key
IATRAKWUXMZMIY-UHFFFAOYSA-N
SMILES
[O--].[Sr++]
Total Metal Impurities
0.5% max.

Description

Strontium oxide was used to make television picture tube glass. It finds application in glass, optic and ceramic industry. It is an active component of paint dryers. It is also used to prepare pure strontium up on heating with aluminum under vacuum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text