Thermo Scientific™

GlycanPac™ AXR-1 HPLC Columns

Catalog number: 088259
Thermo Scientific™

GlycanPac™ AXR-1 HPLC Columns

Catalog number: 088259

Get industry-leading, ultra-high resolution of labeled and native glycans for structural characterization and quantification using the Thermo Scientific™ GlycanPac™ AXR-1 LC Columns. This high-performance, silica-based chromatography column achieves mixed-mode separation using a combination of charge, isomer structure, and size. Now there's a column with unique selectivity to analyze biologically important glycans. The GlycanPac AXR-1 column is available in multiple HPLC and UHPLC formats and is compatible with fluorescence or mass spectrometry (MS) detection.

 
Catalog Number
088259
Unit Size
Pack of 2
Particle Size
3 μm
Diameter (Metric)
3 mm
Length (Metric)
10 mm
Price (USD)
233.00
Your Price:
Availability
***
Full specifications
Quantity
Column FormatGuard Cartridge
Diameter (Metric)3 mm
For Use WithThermo Scientific GlycanPac AXR-1 Column; 3 µm, 3.0 x 150 mm
Length (Metric)10 mm
Max. Pressure6000 psi (415 bar)
Particle Size3 μm
pH2 to 8
Pore Size175 Å
Surface Area300 m²/g
Temperature60°C
USP TypeN/A
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSilica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeGuard Cartridge
Unit SizePack of 2
Catalog Number
Unit Size
Each
Particle Size
1.9 μm
Diameter (Metric)
2.1 mm
Length (Metric)
150 mm
Price (USD)
1,208.00
Your Price:
Availability
***
Full specifications
Quantity
Column ChemistryWAX and RP Mixed Mode
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 to 0.4 mL/min
Length (Metric)150 mm
Max. Pressure10000 psi (690 bar)
Particle ShapeSpherical, high purity, porous
Particle Size1.9 μm
pH2 to 8
Pore Size175 Å
SolventsCompatible with 0 to 100% aqueous and common HPLC solvents
Surface Area220 m²/g
Temperature60°C
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeAnalyticalColumn
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
1.9 μm
Diameter (Metric)
2.1 mm
Length (Metric)
250 mm
Price (USD)
1,266.00
Your Price:
Availability
***
Full specifications
Quantity
Column ChemistryWAX and RP Mixed Mode
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 to 0.4 mL/min
Length (Metric)250 mm
Max. Pressure15000 psi (1035 bar)
Particle ShapeSpherical, high purity, porous
Particle Size1.9 μm
pH2 to 8
Pore Size175 Å
SolventsCompatible with 0 to 100% aqueous and common HPLC solvents
Surface Area220 m²/g
Temperature60°C
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeAnalyticalColumn
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
3 μm
Diameter (Metric)
2.1 mm
Length (Metric)
150 mm
Price (USD)
1,000.00
Your Price:
Availability
***
Full specifications
Quantity
Column ChemistryWAX and RP Mixed Mode
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 to 0.3 mL/min
Length (Metric)150 mm
Max. Pressure6000 psi (415 bar)
Particle ShapeSpherical, high purity, porous
Particle Size3 μm
pH2 to 8
Pore Size175 Å
SolventsCompatible with 0 to 100% aqueous and common HPLC solvents
Surface Area300 m²/g
Temperature60°C
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeAnalyticalColumn
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
3 μm
Diameter (Metric)
3 mm
Length (Metric)
150 mm
Price (USD)
1,000.00
Your Price:
Availability
***
Full specifications
Quantity
Column ChemistryWAX and RP Mixed Mode
Column FormatAnalytical Column
Diameter (Metric)3 mm
Flow Rate0.4 to 0.6 mL/min
Length (Metric)150 mm
Max. Pressure6000 psi (415 bar)
Particle ShapeSpherical, high purity, porous
Particle Size3 μm
pH2 to 8
Pore Size175 Å
SolventsCompatible with 0 to 100% aqueous and common HPLC solvents
Surface Area300 m²/g
Temperature60°C
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
3 μm
Diameter (Metric)
4.6 mm
Length (Metric)
150 mm
Price (USD)
1,000.00
Your Price:
Availability
***
Full specifications
Quantity
Column ChemistryWAX and RP Mixed Mode
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Flow Rate0.8 to 1.2 mL/min
Length (Metric)150 mm
Max. Pressure6000 psi (415 bar)
Particle ShapeSpherical, high purity, porous
Particle Size3 μm
pH2 to 8
Pore Size175 Å
SolventsCompatible with 0 to 100% aqueous and common HPLC solvents
Surface Area300 m²/g
Temperature60°C
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeAnalyticalColumn
Unit SizeEach
Catalog Number
Unit Size
Pack of 2
Particle Size
3 μm
Diameter (Metric)
4.6 mm
Length (Metric)
10 mm
Price (USD)
233.00
Your Price:
Availability
***
Full specifications
Quantity
Column FormatGuard Cartridge
Diameter (Metric)4.6 mm
For Use WithThermo Scientific GlycanPac AXR-1 Column; 3 µm, 4.6 x 150 mm
Length (Metric)10 mm
Max. Pressure6000 psi (415 bar)
Particle Size3 μm
pH2 to 8
Pore Size175 Å
Surface Area300 m²/g
Temperature60°C
USP TypeN/A
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSilica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeGuardCartridge
Unit SizePack of 2
Catalog Number
Unit Size
Pack of 2
Particle Size
3 μm
Diameter (Metric)
2.1 mm
Length (Metric)
10 mm
Price (USD)
233.00
Your Price:
Availability
***
Full specifications
Quantity
Column FormatGuard Cartridge
Diameter (Metric)2.1 mm
For Use WithThermo Scientific GlycanPac AXR-1 Column; 3 µm, 2.1 x 150 mm
Length (Metric)10 mm
Max. Pressure6000 psi (415 bar)
Particle Size3 μm
pH2 to 8
Pore Size175 Å
Surface Area300 m²/g
Temperature60°C
USP TypeN/A
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSilica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeGuard Cartridge
Unit SizePack of 2
Showing 8 of 8
Catalog NumberSpecificationsUnit SizeParticle SizeDiameter (Metric)Length (Metric)Price (USD)AvailabilityQuantity
088259Full specifications
Pack of 23 μm3 mm10 mm
233.00
Your Price:
***
Column FormatGuard Cartridge
Diameter (Metric)3 mm
For Use WithThermo Scientific GlycanPac AXR-1 Column; 3 µm, 3.0 x 150 mm
Length (Metric)10 mm
Max. Pressure6000 psi (415 bar)
Particle Size3 μm
pH2 to 8
Pore Size175 Å
Surface Area300 m²/g
Temperature60°C
USP TypeN/A
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSilica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeGuard Cartridge
Unit SizePack of 2
088136Full specifications
Each1.9 μm2.1 mm150 mm
1,208.00
Your Price:
***
Column ChemistryWAX and RP Mixed Mode
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 to 0.4 mL/min
Length (Metric)150 mm
Max. Pressure10000 psi (690 bar)
Particle ShapeSpherical, high purity, porous
Particle Size1.9 μm
pH2 to 8
Pore Size175 Å
SolventsCompatible with 0 to 100% aqueous and common HPLC solvents
Surface Area220 m²/g
Temperature60°C
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeAnalyticalColumn
Unit SizeEach
088135Full specifications
Each1.9 μm2.1 mm250 mm
1,266.00
Your Price:
***
Column ChemistryWAX and RP Mixed Mode
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 to 0.4 mL/min
Length (Metric)250 mm
Max. Pressure15000 psi (1035 bar)
Particle ShapeSpherical, high purity, porous
Particle Size1.9 μm
pH2 to 8
Pore Size175 Å
SolventsCompatible with 0 to 100% aqueous and common HPLC solvents
Surface Area220 m²/g
Temperature60°C
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeAnalyticalColumn
Unit SizeEach
088251Full specifications
Each3 μm2.1 mm150 mm
1,000.00
Your Price:
***
Column ChemistryWAX and RP Mixed Mode
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 to 0.3 mL/min
Length (Metric)150 mm
Max. Pressure6000 psi (415 bar)
Particle ShapeSpherical, high purity, porous
Particle Size3 μm
pH2 to 8
Pore Size175 Å
SolventsCompatible with 0 to 100% aqueous and common HPLC solvents
Surface Area300 m²/g
Temperature60°C
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeAnalyticalColumn
Unit SizeEach
088252Full specifications
Each3 μm3 mm150 mm
1,000.00
Your Price:
***
Column ChemistryWAX and RP Mixed Mode
Column FormatAnalytical Column
Diameter (Metric)3 mm
Flow Rate0.4 to 0.6 mL/min
Length (Metric)150 mm
Max. Pressure6000 psi (415 bar)
Particle ShapeSpherical, high purity, porous
Particle Size3 μm
pH2 to 8
Pore Size175 Å
SolventsCompatible with 0 to 100% aqueous and common HPLC solvents
Surface Area300 m²/g
Temperature60°C
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeAnalytical Column
Unit SizeEach
088255Full specifications
Each3 μm4.6 mm150 mm
1,000.00
Your Price:
***
Column ChemistryWAX and RP Mixed Mode
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Flow Rate0.8 to 1.2 mL/min
Length (Metric)150 mm
Max. Pressure6000 psi (415 bar)
Particle ShapeSpherical, high purity, porous
Particle Size3 μm
pH2 to 8
Pore Size175 Å
SolventsCompatible with 0 to 100% aqueous and common HPLC solvents
Surface Area300 m²/g
Temperature60°C
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeAnalyticalColumn
Unit SizeEach
088260Full specifications
Pack of 23 μm4.6 mm10 mm
233.00
Your Price:
***
Column FormatGuard Cartridge
Diameter (Metric)4.6 mm
For Use WithThermo Scientific GlycanPac AXR-1 Column; 3 µm, 4.6 x 150 mm
Length (Metric)10 mm
Max. Pressure6000 psi (415 bar)
Particle Size3 μm
pH2 to 8
Pore Size175 Å
Surface Area300 m²/g
Temperature60°C
USP TypeN/A
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSilica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeGuardCartridge
Unit SizePack of 2
088258Full specifications
Pack of 23 μm2.1 mm10 mm
233.00
Your Price:
***
Column FormatGuard Cartridge
Diameter (Metric)2.1 mm
For Use WithThermo Scientific GlycanPac AXR-1 Column; 3 µm, 2.1 x 150 mm
Length (Metric)10 mm
Max. Pressure6000 psi (415 bar)
Particle Size3 μm
pH2 to 8
Pore Size175 Å
Surface Area300 m²/g
Temperature60°C
USP TypeN/A
Stationary PhaseProprietary
Product LineGlycanPac
Packing MaterialSilica
Column TypeMixed Mode
PhaseReversed Phase and Ion Exchange
TypeGuard Cartridge
Unit SizePack of 2
Showing 8 of 8
Exceptional Glycan Selectivity with Industry-Leading Resolution

Ultra-high resolution and selectivity of glycans are attributed to the GlycanPac AXR-1 column's innovative mixed-mode surface chemistry, which combines:

  • WAX functionality for retention of negatively charged glycans, with separation in order of increasing charge
  • Reversed-phase functionality for same-charge glycans, with separation according to isomerism and size
Reliable Column Performance

This high-purity spherical silica column is designed and tested for HPLC and UHPLC methods using either fluorescence or MS detection applications. The GlycanPac AXR-1 column uses volatile aqueous buffers (e.g., ammonium acetate or ammonium formate) and acetonitrile, presenting the eluting glycans ready for introduction into MS instruments.

Available in Multiple Formats
  • Use 1.9 µm columns for UHPLC applications and 3 µm columns for HPLC applications
  • Select from a variety of dimensions to support the resolution, throughput, or instrumental requirements required for your application
  • Protect the analytical column with GycanPac AXR-1 Guard columns
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Figures

Frequently asked questions (FAQs)

Citations & References