Iron nickel oxide, tech.
Iron nickel oxide, tech.
Iron nickel oxide, tech.
Thermo Scientific Chemicals

Iron nickel oxide, tech.

CAS: 12168-54-6 | Fe2NiO4 | 234.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 088530.18
also known as 088530-18
Price (EUR)
218,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
218,00
Each
Chemical Identifiers
CAS12168-54-6
IUPAC Nameoxo[(oxoferrio)oxy]iron; oxonickel
Molecular FormulaFe2NiO4
InChI KeyNQNBVCBUOCNRFZ-UHFFFAOYSA-N
SMILESO=[Ni].O=[Fe]O[Fe]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity content1% max.
Iron nickel oxide is used as an adsorbent to remove hexavalent chromium ions present in the waste water by co-sedimentation method. It is also employed as an additive in membrane (i.e. polyvinylchloride/styrene-butadiene-rubber blend heterogeneous cation exchange membrane) fabrication, thereby increasing the permeability for bivalent ions compared to the monovalent type.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only