Potassium bromate, Acculute Standard Volumetric Solution, Final Concentration 0.1N - FAQs
View additional product information for Potassium bromate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, Thermo Scientific Chemicals - FAQs (088592.LQ)
2 product FAQs found
What is the protocol for diluting one unit of Potassium bromate, Acculute Standard Volumetric Solution, Final Concentration 0.1N (Cat. No. 088592.LQ, 088592-LQ)?
The following is the dilution protocol:
- Transfer entire contents of one unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask. Repeat rinse three times.
- Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 0.1N Potassium bromate.
What is one unit of Potassium bromate, Acculute Standard Volumetric Solution, Final Concentration 0.1N (Cat. No. 088592.LQ, 088592-LQ)?
1 unit of Potassium bromate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, when diluted to 1 L, will yield a final concentration of 0.1N Potassium bromate solution.