Silver nitrate, Acculute Standard Volumetric Solution, Final Concentration 0.1N - FAQs
View additional product information for Silver nitrate, Acculute Standard Volumetric Solution, Final Concentration 0.1N - FAQs (088614.LS, 088614.LQ)
2 product FAQs found
How do I prepare 1 L of 0.1 N Silver nitrate from one unit of Silver nitrate, Acculute Standard Volumetric Solution, Final Concentration 0.1 N?
We recommend the following protocol to prepare 1 L of .1N Silver nitrate:
  1. Transfer the entire contents of the bottle into a 1 L volumetric flask.
  2. Rinse the bottle residue, the threads of the bottle, and the inner bottle cap with deionized water, and transfer to the 1 L flask.
  3. Repeat the step 2 rinse three times.
  4. Bring the 1 L flask to volume with deionized water and mix well. The final solution in the 1 L flask has a concentration of 0.1 N.
What is the size of one unit of Silver nitrate, Acculute Standard Volumetric Solution, Final Concentration 0.1 N?
One unit of Silver nitrate, Acculute Standard Volumetric Solution is 150 mL. When you dilute the unit to 1 L, the final concentration will be 0.1000 N ± 0.0002 N.