Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1L - FAQs
View additional product information for Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1L - FAQs (088632.LQ, 088632.LS)
2 product FAQs found
What are the instructions for dilution of 1 unit of Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1L (Cat. No. 088632.LQ, 088632.LS)?
The following is the dilution protocol:
- Transfer entire contents of 1 unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask.
- Repeat rinse three times. Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 1% Starch Indicator.
What is one unit of Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1L?
One unit of Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1 L makes one liter of 1% solution of starch indicator after dilution to one liter.