Search
Search

Use the new Thermo Scientific™ MAbPac™ RP columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins.
| Número do catálogo | Length (Metric) | Diameter (Metric) | Stationary Phase |
|---|---|---|---|
| 088648 | 50 mm | 2.1 mm | Phenyl (Ph) |
| 088644 | 100 mm | 3 mm | Phenyl (Ph) |
| 088645 | 50 mm | 3 mm | Phenyl (Ph) |
| 088646 | 10 mm | 3 mm | Phenyl (Ph) |
| 088647 | 100 mm | 2.1 mm | Phenyl (Ph) |
| 088649 | 10 mm | 2.1 mm | Phenyl (Ph) |
| 303182 | 50 mm | 1 mm | PSDVB (Polystyrene Divinylbenzene) |
| 303183 | 100 mm | 1 mm | PSDVB (Polystyrene Divinylbenzene) |
| 303184 | 150 mm | 1 mm | PSDVB (Polystyrene Divinylbenzene) |
| 303269 | 150 mm | 3 mm | Polystyrene-divinylbenzene (PS-DVB) |
| 303270 | 150 mm | 2.1 mm | Polystyrene-Divinylbenzene (PS-DVB) |
Use the new Thermo Scientific™ MAbPac™ Reversed Phase columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over. The surface chemistry, particle size, and pore size, have been optimized for high resolution separations.
Compatible with organic solvent used in HPLC and MS under RP conditions
Efficient with low carry over
Designed for reversed-phase biomolecule LC and LC/MS separations