Dionex™ DRS 600 Dynamically Regenerated Suppressor
Dionex™ DRS 600 Dynamically Regenerated Suppressor
Actual product may vary
Dionex™ DRS 600 Dynamically Regenerated Suppressor
Dionex™ DRS 600 Dynamically Regenerated Suppressor
Thermo Scientific™

Dionex™ DRS 600 Dynamically Regenerated Suppressor

Please note these suppressors will be discontinued on December 31st, 2025. The recommended replacement is the Thermo Scientific™ Dionex™ NGES Next Generation Electrolytic Suppressor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFormat
088670Cation Dynamically Regenerated Suppressor2 mm
088666Anion Dynamically Regenerated Suppressor4 mm
088667Anion Dynamically Regenerated Suppressor2 mm
Catalog number 088670
Price (MXN)
-
Request A Quote
Type:
Cation Dynamically Regenerated Suppressor
Format:
2 mm

Please note: The Thermo Scientific™ Dionex™ DRS 600 suppressors will be discontinued on December 31st, 2025. The Dionex NGES Next Generation Electrolytic Suppressor is the recommended replacement. For more information, please visit the Dionex NGES Electrolytic Suppressor page.

Use the Thermo Scientific™ Dionex™ DRS 600 Dynamically Regenerated Suppressor for electrolytically regenerated suppression in ion chromatography (IC) applications requiring high capacity, low noise, high backpressure resiliency, and fast startup. This suppressor is a fundamental component of a reagent-free ion chromatography (RFIC™) system.

Key features

  • Full electrolytic regeneration without reagents
  • Minimizes training and operation time by eliminating error-prone manual calculation of electrical currents
  • Improves gradient performance by improving signal-to-noise ratio with gradient eluents by dynamically adapting to the changing eluent composition
  • Compatible across a wide variety of applications, including IC coupled with mass spectrometry (IC-MS)

The Dionex DRS 600 Dynamically Regenerated Suppressor is the recommended replacement for the Thermo Scientific™ Dionex™ ERS™ 500 Electrolytically Regenerated Suppressor.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for six (6) months thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Amperage110 mA
DescriptionReplacement for P/N 082543 Dionex CERS Cation Electrolytically Regenerated Suppressor (2 mm)
Flow Rate0.75 mL/min. Max.
For Use With (Application)Ion chromatography applications requiring high capacity, low noise, high backpressure resiliency, and fast startup
Format2 mm
OperationUsed for electrolytically regenerated suppression in ion chromatography.
Pressure30 to 100 psi (recommended)
Product LineDionex CDRS 600
Temperature Range15°C to 50°C
TypeCation Dynamically Regenerated Suppressor
Unit SizeEach 1