Ethylenediaminetetraacetic acid, iron(III) monosodium salt
Ethylenediaminetetraacetic acid, iron(III) monosodium salt
Ethylenediaminetetraacetic acid, iron(III) monosodium salt
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, iron(III) monosodium salt

CAS: 15708-41-5 | C10H12FeN2NaO8 | 367.047 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 088995.22
also known as 088995-22
Price (USD)
46.60
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
46.60
Each
Chemical Identifiers
CAS15708-41-5
IUPAC Nameiron(3+) sodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Molecular FormulaC10H12FeN2NaO8
InChI KeyMKWYFZFMAMBPQK-UHFFFAOYSA-J
SMILES[Na+].[Fe+3].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)Total EDTA: ≥65.5%
Formpowder
Appearance (Color)Pale yellow
Assay (Titration)Fe (III) 12.5 - 13.5%
Ethylenediaminetetraacetic acid, iron(III) monosodium salt is used to eliminate enzyme inhibition by traces of heavy metals and to inhibit enzymes that require divalent cations as cofactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only