Hidruro de titanio(II), 99 % (base metálica), Thermo Scientific Chemicals
Hidruro de titanio(II), 99 % (base metálica), Thermo Scientific Chemicals
Hidruro de titanio(II), 99 % (base metálica), Thermo Scientific Chemicals
Hidruro de titanio(II), 99 % (base metálica), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hidruro de titanio(II), 99 % (base metálica), Thermo Scientific Chemicals

CAS: 7704-98-5 | H2Ti
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
250 g
Número de catálogo 089183.30
también denominado 089183-30
Precio (USD)
-
Cantidad:
250 g
Pedido a granel o personalizado
Identificadores químicos
CAS7704-98-5
IUPAC Nametitanium(2+) dihydride
Molecular FormulaH2Ti
InChI KeyKAZWGWWZKAHTKC-UHFFFAOYSA-N
SMILES[H-].[H-].[Ti++]
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Assay from Suppliers CofAHydrogen, H: ≥3.7%
Assay from Suppliers CofATitanium, Ti: ≥94.0% (total Ti)
Gain on Ignition≥56.7%
Particle Size≥99.9% is <45µm
Particle SizeAverage: 1.6-2.0µm
Ver más
For preparation of titanium compounds and coatings, in formation of glass and ceramic seals, as a catalyst, for thermal storage, as a source for pure H{2}Titanium(II) hydride is widely utilized as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. It is involved in various applications such as ceramics, pyrotechnics and sports equipment. It is also used as a laboratory reagent and as a blowing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only