Sodium hydroxide solution, 50%, WFI, Biotech Reagent, GMP, J.T.Baker™
Sodium hydroxide solution, 50%, WFI, Biotech Reagent, GMP, J.T.Baker™
Avantor™

Sodium hydroxide solution, 50%, WFI, Biotech Reagent, GMP, J.T.Baker™

Made from Water for Injection (WFI) U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial). Meets BP, Ph.Eur. and JP Chemical specifications.Read more
Have Questions?
Change viewbuttonViewtableView
Quantity:
19 L
200 L
Catalog number 0897-07
Price (MXN)
-
Request A Quote
Quantity:
19 L
Made from Water for Injection (WFI) U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial). Meets BP, Ph.Eur. and JP Chemical specifications.
Specifications
Boiling Point119°C to 135°C (1013 hPa)
CAS1310-73-2
Chemical Name or MaterialSodium hydroxide
ColorColorless
Melting Point9°C to 10°C
Molecular FormulaNaOH
PackagingPoly bottle
SynonymSodium hydrate, Soda lye, Caustic soda, Lye
Molecular Weight (g/mol)40
Quantity19 L
Unit SizeEach