Sodium hydroxide solution, 50%, WFI, Biotech Reagent, GMP, J.T.Baker™
Avantor™

Sodium hydroxide solution, 50%, WFI, Biotech Reagent, GMP, J.T.Baker™

Made from Water for Injection (WFI) U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial). Meets BP, Ph.Eur. and JP Chemical specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
19 L
200 L
Número de catálogo 0897-09
Precio (MXN)
-
Cantidad:
200 L
Made from Water for Injection (WFI) U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial). Meets BP, Ph.Eur. and JP Chemical specifications.
Made from Water for Injection (WFI) U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial). Meets BP, Ph.Eur. and JP Chemical specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Punto de ebullición119°C to 135°C (1013 hPa)
CAS1310-73-2
Nombre del producto químico o materialSodium hydroxide
ColorColorless
Punto de fusión9°C to 10°C
Fórmula molecularNaOH
EnvasePolyethylene drum
SinónimoSodium hydrate, Soda lye, Caustic soda, Lye
GradoBiotech
Peso molecular (g/mol)40
Cantidad200 L
Unit Size200 L