Thermo Scientific Chemicals

Calcium phosphate (pyro), 96% min, Thermo Scientific Chemicals

Catalog number: 089836.30
250 g, Each
Thermo Scientific Chemicals

Calcium phosphate (pyro), 96% min, Thermo Scientific Chemicals

Catalog number: 089836.30
250 g, Each
Quantity
Catalog number: 089836.30
also known as 089836-30
Price (USD)
Price: 36.40
Online price: 31.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7790-76-3
IUPAC Name
dicalcium (phosphonatooxy)phosphonate
Molecular Formula
Ca2O7P2
InChI Key
JUNWLZAGQLJVLR-UHFFFAOYSA-J
SMILES
[Ca++].[Ca++].[O-]P([O-])(=O)OP([O-])([O-])=O
Assay (unspecified)
96% min

Description

Calcium phosphate is used in the production of phosphoric acid and fertilizers. It is used in baking as the acid in a leavening agent. It is also used in cheese products. This compound has also been used in gene transfection of cells. It is also being used in the development of contrast agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text