Oxone∣r, monopersulfate
Oxone∣r, monopersulfate
Oxone∣r, monopersulfate
Thermo Scientific Chemicals

Oxone∣r, monopersulfate

CAS: 70693-62-8 | H3K5O18S4 | 614.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
Catalog number 089892.A1
also known as 089892-A1
Price (USD)
77.00
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
77.00
Each
Chemical Identifiers
CAS70693-62-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormGranular free-flowing powder
Assay≥4.5% [active oxygen]
Assay (unspecified)≥4.5% [active oxygen]
Residual water≤0.1% (≤1000ppm)
Oxone is used for halogenation of a,b-unsaturated carbonyl compounds and catalytic generation of hypervalent iodine reagents for alcohol oxidation. It is a used for rapid, and good synthesis of oxaziridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only