Thermo Scientific Chemicals

Strontium sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 089895.18
50 g, Each
Thermo Scientific Chemicals

Strontium sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 089895.18
50 g, Each
Quantity
Catalog number: 089895.18
also known as 089895-18
Price (USD)
Price: 557.00
Online price: 482.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1314-96-1
IUPAC Name
strontium(2+) sulfanediide
Molecular Formula
SSr
InChI Key
ZEGFMFQPWDMMEP-UHFFFAOYSA-N
SMILES
[S--].[Sr++]
Total Metal Impurities
0.1% max.

Description

Strontium sulfide is used in chemical research and as organic intermediates. It is also used as depilatory. It is also used as Isotope labelled enantiomer of Voriconazole (V760000), an antifungal agent and an ergosterol biosynthesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text