Pretreatment Cartridges and Filters
Pretreatment Cartridges and Filters
Pretreatment Cartridges and Filters
Thermo Scientific™
Pretreatment Cartridges and Filters
Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Have Questions?
變更視圖buttonViewtableView
產品號碼Type
09.4000Pretreatment Cartridge
D8830Prefilter MQ Cartridge
D63111Prefilter
06.5204Filter with Stabilizer
06.5452Stabilizing Cartridge
09.4003
亦稱為 9.4003
Pretreatment Filter
50134022Filter
產品號碼 09.4000
價格 (HKD)
-
Type:
Pretreatment Cartridge
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.
規格
Includes5 μm Filter w/ Carbon Cartridge, Hardness Stabilizer Cartridge
For Use With (Equipment)LabTower TII, Pacific RO, Pacific TII
TypePretreatment Cartridge
Unit SizeEach