Polyethylene Glove Bag, 43cm (17in) x 43cm (17in) x 28cm (11in), 29cm (11.5in) entrance - FAQs
View additional product information for Polyethylene Glove Bag, 43cm (17in) x 43cm (17in) x 28cm (11in), 29cm (11.5in) entrance - FAQs (091717.LL)
1 product FAQs found
What comes with the Polyethylene Glove Bag, 43cm (17in) x 43cm (17in) x 28cm (11in), 29cm (11.5in) entrance (Cat. No. 091717.LL)?
Polyethylene Glove Bag, 43cm (17in) x 43cm (17in) x 28cm (11in), 29cm (11.5in) entrance comes with the following:
  • 4 clips for closure
  • A 6-inch clear tube
  • 1 connector reducer
  • A pair of cotton gloves