Thermo Scientific™
Dionex™ ICS-Series PDA Photodiode Array Detector Replacement Lamps
Catalog number: 0939016T
Thermo Scientific™
Dionex™ ICS-Series PDA Photodiode Array Detector Replacement Lamps
Catalog number: 0939016T
Thermo Scientific™ Dionex™ Replacement Photodiode Array Detector Lamps encompass the wavelength range from 190 to 800 nm, providing greater applications flexibility.
Catalog Number
0939016T
Unit Size
Each
Type
Deuterium Lamp, 190-380 nm
Price (USD)
Full specifications
For Use WithPhotodiode Array Detectors
OperationEnsure low baseline noise, high signal intensity, and applications flexibility.
TypeDeuterium Lamp, 190-380 nm
Detector TypeUV
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Tungsten Lamp, 380-800nm
Price (USD)
Full specifications
For Use WithPhotodiode Array Detectors
OperationEnsure low baseline noise, high signal intensity, and applications flexibility.
TypeTungsten Lamp, 380-800nm
Detector TypeVisible
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeTypePrice (USD)
0939016TFull specifications
EachDeuterium Lamp, 190-380 nmRequest A Quote
For Use WithPhotodiode Array Detectors
OperationEnsure low baseline noise, high signal intensity, and applications flexibility.
TypeDeuterium Lamp, 190-380 nm
Detector TypeUV
Unit SizeEach
056123TFull specifications
EachTungsten Lamp, 380-800nmRequest A Quote
For Use WithPhotodiode Array Detectors
OperationEnsure low baseline noise, high signal intensity, and applications flexibility.
TypeTungsten Lamp, 380-800nm
Detector TypeVisible
Unit SizeEach
Showing 2 of 2
  • The deuterium and tungsten lamps ensure low baseline noise, high signal intensity and applications flexibility
  • Deuterium lamp optimizes the UV range (190 to 380 nm)
  • Tungsten lamp optimizes the visible range (380 to 800 nm)