Are replacement Imhoff cone closures available?
Yes, we do offer replacements. Please contact Thermo Scientific Laboratory Products Essentials technical support for more information.
Search
View additional product information for Nalgene™ Polycarbonate Imhoff Settling Cone - FAQs (1000-0010PK)
1 product FAQs found