Product Image
Thermo Scientific™

CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators

Save time while maintaining accuracy with economical Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators, optimized for use with CEDIA TDM Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionDetectable Analytes
100001TS
also known as 100001
CEDIA Cardiac TDM Multi-CalibratorsNAPA, Procainamide
100007CEDIA Antiepileptic TDM Multi-CalibratorsCarbamazepine, Phenobarbital, Phenytoin, Theophylline, Valproic Acid
100017CEDIA Antibiotic TDM Multi-CalibratorsGentamicin, Tobramycin
Catalog number 100001TS
also known as 100001
Price (USD)
-
Description:
CEDIA Cardiac TDM Multi-Calibrators
Detectable Analytes:
NAPA, Procainamide
Streamline testing with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators, a single calibrator kit that can be used with multiple CEDIA TDM assays. These economical kits are valued for their accuracy, precision and lot-to-lot dependability. They are available for cardiac or antiarrhythmic, antiepileptic and antibiotic drug monitoring in human serum and plasma. Each kit includes both high and low calibrators.
  • Proven technology
  • Liquid ready-to-use: no reconstitution needed
  • Multiple drug constituents per calibration kit
  • Positive linear calibration curve: the higher the absorbance reading, the more drug present
  • Excellent correlation to reference methods
  • Recombinant DNA technology provides lot-to-lot consistency
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionCEDIA Cardiac TDM Multi-Calibrators
QuantityLow: 2 x 7.5 mL; High: 2 x 5.0 mL
TypeCalibrator
Detectable AnalytesNAPA, Procainamide
Sample TypePlasma, Serum
Unit SizeEach
Contents & Storage
2°C to 8°C