CEDIA™ Cyclosporine (CsA) Immunoassays and Calibrators
Actual product may vary
CEDIA™ Cyclosporine (CsA) Immunoassays and Calibrators
CEDIA™ Cyclosporine (CsA) Immunoassays and Calibrators
Thermo Scientific™
CEDIA™ Cyclosporine (CsA) Immunoassays and Calibrators
Achieve optimal low-end sensitivity and a broad dynamic range with the Thermo Scientific™ CEDIA™ Cyclosporine Plus Immunoassay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
100012CsA PLUS High Calibrator Set
100147CsA PLUS Combo Kit
Catalog number 100012
Price (MXN)
-
Type:
CsA PLUS High Calibrator Set
Achieve precise and accurate quantitative results crucial for monitoring drug levels of cyclosporine in kidney, liver and heart transplant patients with our easy-to-use Thermo Scientific™ CEDIA™ Cyclosporine Plus Immunoassay. Used with the Thermo Scientific CEDIA Plus High Range Calibrators, this immunoassay achieves a broad dynamic range of 25-2000ng/mL for both trough and peak samples. Applications are available on our easy-to-use Indiko™ and Indiko Plus clinical and specialty chemistry systems as well as a wide variety of other laboratory instrumentation.
Key Features
  • Optimal low-end sensitivity with a broad dynamic range
  • Excellent correlation with HPLC-MS, the gold standard method
  • Precise throughout the standard curve assay range
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Quantity4 x 2 mL each level, low and high
Sample TypePlasma, Serum
TypeCsA PLUS High Calibrator Set
Detectable AnalytesCyclosporin A
Unit SizeEach
Contents & Storage
2°C to 8°C